Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCCTCTTCTTTTTCTTCCCATT[A/G]GGCATAACATCTCATTCACCCCAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616904 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOCK5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DOCK5 - dedicator of cytokinesis 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322810.1 | Intron | NP_001309739.1 | ||||
NM_024940.7 | Intron | NP_079216.4 | ||||
XM_011544661.1 | Intron | XP_011542963.1 | ||||
XM_011544662.1 | Intron | XP_011542964.1 | ||||
XM_011544663.2 | Intron | XP_011542965.1 | ||||
XM_017013865.1 | Intron | XP_016869354.1 | ||||
XM_017013866.1 | Intron | XP_016869355.1 | ||||
XM_017013867.1 | Intron | XP_016869356.1 | ||||
XM_017013868.1 | Intron | XP_016869357.1 |