Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCTTCTGGGTTGCATACATCCGG[C/G]TCTCCATGGGGCAGTCCTGACTCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 193002 | ||||||||||||||||||||
Literature Links: |
SLC18A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC18A1 - solute carrier family 18 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135691.2 | 1562 | Missense Mutation | ACC,AGC | T,S 499 | NP_001129163.1 | |
NM_001142324.1 | 1562 | Missense Mutation | ACC,AGC | T,S 467 | NP_001135796.1 | |
NM_001142325.1 | 1562 | Silent Mutation | GAC,GAG | D,E 454 | NP_001135797.1 | |
NM_003053.3 | 1562 | Missense Mutation | ACC,AGC | T,S 499 | NP_003044.1 | |
XM_011544625.1 | 1562 | Missense Mutation | ACC,AGC | T,S 471 | XP_011542927.1 | |
XM_011544626.1 | 1562 | Missense Mutation | ACC,AGC | T,S 467 | XP_011542928.1 | |
XM_011544627.1 | 1562 | Missense Mutation | GAC,GAG | D,E 454 | XP_011542929.1 |