Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAATGCTATTTTTTATGCATTAT[A/T]ATAATTTTGTAAATAAGAAATGTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 170993 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PEX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PEX2 - peroxisomal biogenesis factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000318.2 | 2977 | UTR 3 | NP_000309.1 | |||
NM_001079867.1 | 2977 | UTR 3 | NP_001073336.1 | |||
NM_001172086.1 | 2977 | UTR 3 | NP_001165557.1 | |||
NM_001172087.1 | 2977 | UTR 3 | NP_001165558.1 |