Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTATTATATAACTATATACTTAA[C/T]ACATCAAATTGTAACCTTGTATGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603912 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF3H PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF3H - eukaryotic translation initiation factor 3 subunit H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003756.2 | Intron | NP_003747.1 |
LOC105375713 - uncharacterized LOC105375713 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |