Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCACTTGCACACATTTTATTGTAC[A/G]AAATTCCCCAGTACTGCTTTAATAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606387 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOP1MT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TOP1MT - topoisomerase (DNA) I, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258446.1 | 1910 | UTR 3 | NP_001245375.1 | |||
NM_001258447.1 | 1910 | UTR 3 | NP_001245376.1 | |||
NM_052963.2 | 1910 | UTR 3 | NP_443195.1 | |||
XM_005250781.3 | 1910 | Intron | XP_005250838.1 |
ZNF696 - zinc finger protein 696 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |