Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGAACAATGTATTAATATCTTAT[A/G]TGGAAATTGCTAACTGTTCCCTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIQK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRIQK - triple QxxK/R motif containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171795.1 | Intron | NP_001165266.1 | ||||
NM_001171796.1 | Intron | NP_001165267.1 | ||||
NM_001171797.1 | Intron | NP_001165268.1 | ||||
NM_001171798.1 | Intron | NP_001165269.1 | ||||
NM_001171799.1 | Intron | NP_001165270.1 | ||||
NM_001191035.1 | Intron | NP_001177964.1 | ||||
NM_001191036.1 | Intron | NP_001177965.1 | ||||
XM_017013347.1 | Intron | XP_016868836.1 | ||||
XM_017013348.1 | Intron | XP_016868837.1 | ||||
XM_017013349.1 | Intron | XP_016868838.1 | ||||
XM_017013350.1 | Intron | XP_016868839.1 |