Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATACAAGGCAATTGAAGGCATTCA[A/G]TAATGTAAGTGTTATCATTGGGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611297 MIM: 605030 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OSR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OSR2 - odd-skipped related transciption factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142462.2 | Intron | NP_001135934.1 | ||||
NM_001286841.1 | Intron | NP_001273770.1 | ||||
NM_053001.3 | Intron | NP_443727.2 | ||||
XM_005250778.4 | Intron | XP_005250835.1 | ||||
XM_011516825.2 | Intron | XP_011515127.1 | ||||
XM_011516826.2 | Intron | XP_011515128.1 | ||||
XM_011516827.2 | Intron | XP_011515129.1 | ||||
XM_017013018.1 | Intron | XP_016868507.1 |
STK3 - serine/threonine kinase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |