Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCAAAGTACATCAAATGAGATTA[A/G]GAATAATGTAATGATTTTCTTTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 132811 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPHX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPHX2 - epoxide hydrolase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256482.1 | Intron | NP_001243411.1 | ||||
NM_001256483.1 | Intron | NP_001243412.1 | ||||
NM_001256484.1 | Intron | NP_001243413.1 | ||||
NM_001979.5 | Intron | NP_001970.2 | ||||
XM_017013199.1 | Intron | XP_016868688.1 | ||||
XM_017013200.1 | Intron | XP_016868689.1 |