Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAAGTTGAGATCAATGCCTGTCT[A/G]TCCTACAGGATCTATCCTATGGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611605 MIM: 604436 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERLIN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERLIN2 - ER lipid raft associated 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PROSC - proline synthetase cotranscribed homolog (bacterial) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007198.3 | Intron | NP_009129.1 | ||||
XM_017013001.1 | Intron | XP_016868490.1 |