Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGACGTGTATTTTTCTACCCCTC[C/G]TAAGAGGGCACAGTCTCTTCACCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603409 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FZD6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FZD6 - frizzled class receptor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164615.1 | 363 | Missense Mutation | CTA,GTA | L,V 16 | NP_001158087.1 | |
NM_001164616.1 | 363 | Intron | NP_001158088.1 | |||
NM_001317796.1 | 363 | UTR 5 | NP_001304725.1 | |||
NM_003506.3 | 363 | Missense Mutation | CTA,GTA | L,V 16 | NP_003497.2 |