Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAACTCGGGAACCCCATCAAAGGA[C/G]GGTGGCTTCAGGATGACTTCAAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
STMN4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STMN4 - stathmin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001283053.1 | 289 | Silent Mutation | CCC,CCG | P,P 98 | NP_001269982.1 | |
NM_001283054.1 | 289 | Silent Mutation | CCC,CCG | P,P 71 | NP_001269983.1 | |
NM_001283055.1 | 289 | Silent Mutation | CCC,CCG | P,P 62 | NP_001269984.1 | |
NM_030795.3 | 289 | Silent Mutation | CCC,CCG | P,P 98 | NP_110422.2 | |
XM_005273652.4 | 289 | Silent Mutation | CCC,CCG | P,P 98 | XP_005273709.1 | |
XM_005273655.4 | 289 | Silent Mutation | CCC,CCG | P,P 71 | XP_005273712.1 |