Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTGTTTCTGTTGTGGTGGGATG[G/T]ATTGCAGTGAATGAATCTTCATCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611798 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFR3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EFR3A - EFR3 homolog A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323553.1 | Intron | NP_001310482.1 | ||||
NM_001323554.1 | Intron | NP_001310483.1 | ||||
NM_001323555.1 | Intron | NP_001310484.1 | ||||
NM_001323556.1 | Intron | NP_001310485.1 | ||||
NM_001323557.1 | Intron | NP_001310486.1 | ||||
NM_001323558.1 | Intron | NP_001310487.1 | ||||
NM_015137.5 | Intron | NP_055952.2 |