Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACTCTCTGCACGCTGGGCAAGGC[C/T]GAGGTGGGAGGCCCTCTGCGCGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM160B2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM160B2 - family with sequence similarity 160 member B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022749.5 | 345 | Silent Mutation | GCC,GCT | A,A 98 | NP_073586.5 | |
XM_005273602.3 | 345 | Silent Mutation | GCC,GCT | A,A 98 | XP_005273659.1 | |
XM_006716381.2 | 345 | Silent Mutation | GCC,GCT | A,A 98 | XP_006716444.1 | |
XM_006716385.3 | 345 | UTR 5 | XP_006716448.1 | |||
XM_017013732.1 | 345 | Silent Mutation | GCC,GCT | A,A 98 | XP_016869221.1 | |
XM_017013733.1 | 345 | Silent Mutation | GCC,GCT | A,A 40 | XP_016869222.1 | |
XM_017013734.1 | 345 | Silent Mutation | GCC,GCT | A,A 40 | XP_016869223.1 |