Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGTGCTCACTTGGGGGAAGGAA[C/T]CTTGCAAACTCTAAGATGCCTTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118502 MIM: 601212 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRNA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHRNA2 - cholinergic receptor nicotinic alpha 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000742.3 | Intron | NP_000733.2 | ||||
NM_001282455.1 | Intron | NP_001269384.1 | ||||
XM_005273397.2 | Intron | XP_005273454.1 | ||||
XM_006716282.1 | Intron | XP_006716345.1 | ||||
XM_011544388.1 | Intron | XP_011542690.1 | ||||
XM_011544389.1 | Intron | XP_011542691.1 | ||||
XM_017013003.1 | Intron | XP_016868492.1 | ||||
XM_017013004.1 | Intron | XP_016868493.1 |
PTK2B - protein tyrosine kinase 2 beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |