Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACCAAGTAAAGATGATAGAACAT[A/G]TTAGATCTGAATTCCTTTTTGTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607591 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C8orf44-SGK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C8orf44-SGK3 - C8orf44-SGK3 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204173.1 | Intron | NP_001191102.1 |
SGK3 - serum/glucocorticoid regulated kinase family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033578.2 | Intron | NP_001028750.1 | ||||
NM_013257.4 | Intron | NP_037389.4 | ||||
NM_170709.2 | Intron | NP_733827.2 |