Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAAGATTAAAATTAAAAAACTGA[C/T]GCAAAGAGAAGTTAATTTGCCCAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600967 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C8orf59 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C8orf59 - chromosome 8 open reading frame 59 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099670.2 | Intron | NP_001093140.1 | ||||
NM_001099671.2 | Intron | NP_001093141.1 | ||||
NM_001099672.2 | Intron | NP_001093142.1 | ||||
NM_001099673.2 | Intron | NP_001093143.1 | ||||
NM_001293320.1 | Intron | NP_001280249.1 | ||||
XM_011517528.2 | Intron | XP_011515830.1 |
E2F5 - E2F transcription factor 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |