Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGTGAAAGTGAAACGGAGCAGCT[C/T]TCTGGGTTATTGGCAGTGCACGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606604 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO32 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO32 - F-box protein 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242463.1 | 888 | Silent Mutation | GAA,GAG | E,E 245 | NP_001229392.1 | |
NM_058229.3 | 888 | Silent Mutation | GAA,GAG | E,E 338 | NP_478136.1 | |
NM_148177.2 | 888 | Silent Mutation | GAA,GAG | E,E 193 | NP_680482.1 |