Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACATAGGCTCTGGGGTCAGGGCC[A/G]ATCTGGAATCTTGACTTAGCCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605953 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASAP1 - ArfGAP with SH3 domain, ankyrin repeat and PH domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001247996.1 | Intron | NP_001234925.1 | ||||
NM_018482.3 | Intron | NP_060952.2 | ||||
XM_005250925.1 | Intron | XP_005250982.1 | ||||
XM_006716563.3 | Intron | XP_006716626.1 | ||||
XM_006716564.1 | Intron | XP_006716627.1 | ||||
XM_006716565.3 | Intron | XP_006716628.1 | ||||
XM_006716566.1 | Intron | XP_006716629.1 | ||||
XM_011517052.2 | Intron | XP_011515354.1 | ||||
XM_011517053.1 | Intron | XP_011515355.1 | ||||
XM_017013467.1 | Intron | XP_016868956.1 | ||||
XM_017013468.1 | Intron | XP_016868957.1 | ||||
XM_017013469.1 | Intron | XP_016868958.1 |