Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCCAGGCTGAGCAGGAGCTGTAC[A/G]CACAGGGCCCAGGGGCTGCTGGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C8orf31 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C8orf31 - chromosome 8 open reading frame 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173687.3 | 501 | Silent Mutation | ACA,ACG | T,T 32 | NP_775958.1 | |
XM_017013344.1 | 501 | Intron | XP_016868833.1 | |||
XM_017013345.1 | 501 | Intron | XP_016868834.1 | |||
XM_017013346.1 | 501 | Intron | XP_016868835.1 |
LOC107984097 - uncharacterized protein FLJ40521 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |