Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCACCAAAGATTATCCATCATT[A/G]TGGGGCTTTGGAACAACAAAAACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606033 MIM: 603395 | ||||||||||||||||||||
Literature Links: |
POLR2K PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POLR2K - RNA polymerase II subunit K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPAG1 - sperm associated antigen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003114.4 | 113 | Silent Mutation | TTA,TTG | L,L 9 | NP_003105.2 | |
NM_172218.2 | 113 | Silent Mutation | TTA,TTG | L,L 9 | NP_757367.1 | |
XM_011517240.2 | 113 | Silent Mutation | TTA,TTG | L,L 9 | XP_011515542.1 | |
XM_011517241.2 | 113 | Silent Mutation | TTA,TTG | L,L 9 | XP_011515543.1 | |
XM_011517242.2 | 113 | Silent Mutation | TTA,TTG | L,L 9 | XP_011515544.1 | |
XM_011517243.2 | 113 | Silent Mutation | TTA,TTG | L,L 9 | XP_011515545.1 | |
XM_011517245.2 | 113 | Silent Mutation | TTA,TTG | L,L 9 | XP_011515547.1 | |
XM_017013754.1 | 113 | Intron | XP_016869243.1 | |||
XM_017013755.1 | 113 | Intron | XP_016869244.1 |