Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGATTTCTTAATACTTGGCTTCCT[C/T]TTGGGTTGGTTTTCCTGGACTGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605417 MIM: 174760 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DKK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DKK4 - dickkopf WNT signaling pathway inhibitor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014420.2 | 787 | Silent Mutation | AAA,AAG | K,K 126 | NP_055235.1 | |
XM_011544488.2 | 787 | Silent Mutation | AAA,AAG | K,K 126 | XP_011542790.1 | |
XM_017013316.1 | 787 | Silent Mutation | AAA,AAG | K,K 126 | XP_016868805.1 |
POLB - DNA polymerase beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |