Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGCGGCAGGTGTGTTCGATTTT[C/G]AGCTTGTTGATGTCGCCACGCAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610085 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM167A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM167A - family with sequence similarity 167 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_053279.2 | 1345 | Silent Mutation | CTC,CTG | L,L 151 | NP_444509.2 | |
XM_005272398.4 | 1345 | Silent Mutation | CTC,CTG | L,L 151 | XP_005272455.1 | |
XM_011543837.1 | 1345 | Silent Mutation | CTC,CTG | L,L 151 | XP_011542139.1 | |
XM_011543838.2 | 1345 | Silent Mutation | CTC,CTG | L,L 151 | XP_011542140.1 | |
XM_011543840.2 | 1345 | Silent Mutation | CTC,CTG | L,L 151 | XP_011542142.1 |
FAM167A-AS1 - FAM167A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |