Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGTCAGCACCCAGCACGGATCT[A/G]TCCTGCAGCTGAACGACACCTTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610453 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HGSNAT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HGSNAT - heparan-alpha-glucosaminide N-acetyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152419.2 | 376 | Missense Mutation | ATC,GTC | I,V 110 | NP_689632.2 | |
XM_005273409.1 | 376 | Missense Mutation | ATC,GTC | I,V 110 | XP_005273466.1 | |
XM_005273410.1 | 376 | Missense Mutation | ATC,GTC | I,V 110 | XP_005273467.1 | |
XM_005273411.1 | 376 | Missense Mutation | ATC,GTC | I,V 110 | XP_005273468.1 | |
XM_005273412.3 | 376 | Missense Mutation | ATC,GTC | I,V 110 | XP_005273469.1 | |
XM_017013088.1 | 376 | Missense Mutation | ATC,GTC | I,V 110 | XP_016868577.1 | |
XM_017013089.1 | 376 | UTR 5 | XP_016868578.1 |