Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGCTTGGGATTCTTTGGGTGGTG[G/T]TGGGGAGGTGTGAGCAGAGTCCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610312 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100507071 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100507071 - uncharacterized LOC100507071 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIWIL2 - piwi like RNA-mediated gene silencing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135721.1 | Intron | NP_001129193.1 | ||||
NM_018068.3 | Intron | NP_060538.2 | ||||
XM_005273550.4 | Intron | XP_005273607.1 | ||||
XM_005273551.3 | Intron | XP_005273608.1 |