Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCTTGCATCCTCTGCAAAGGAGA[C/T]GGCAAAGTCCTGGTCATTTGCCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601157 MIM: 600471 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEFA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DEFA4 - defensin alpha 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DEFA6 - defensin alpha 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001926.3 | 192 | Missense Mutation | ATC,GTC | I,V 51 | NP_001917.1 |