Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAAAAAGAGAGGAAAGCATACCA[G/T]AAGTCCCTATGGATTAGTTAAAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601099 MIM: 188450 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR7848 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR7848 - microRNA 7848 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLA - Src-like-adaptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001045556.2 | 1756 | UTR 3 | NP_001039021.1 | |||
NM_001045557.2 | 1756 | UTR 3 | NP_001039022.2 | |||
NM_001282964.1 | 1756 | UTR 3 | NP_001269893.1 | |||
NM_001282965.1 | 1756 | UTR 3 | NP_001269894.1 | |||
NM_006748.3 | 1756 | UTR 3 | NP_006739.2 | |||
XM_017013739.1 | 1756 | UTR 3 | XP_016869228.1 |
TG - thyroglobulin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003235.4 | 1756 | Intron | NP_003226.4 | |||
XM_005251038.4 | 1756 | Intron | XP_005251095.1 | |||
XM_005251040.4 | 1756 | Intron | XP_005251097.1 | |||
XM_005251042.4 | 1756 | Intron | XP_005251099.1 | |||
XM_006716622.3 | 1756 | Intron | XP_006716685.1 | |||
XM_017013793.1 | 1756 | Intron | XP_016869282.1 | |||
XM_017013794.1 | 1756 | Intron | XP_016869283.1 | |||
XM_017013795.1 | 1756 | Intron | XP_016869284.1 | |||
XM_017013796.1 | 1756 | Intron | XP_016869285.1 | |||
XM_017013797.1 | 1756 | Intron | XP_016869286.1 | |||
XM_017013798.1 | 1756 | Intron | XP_016869287.1 | |||
XM_017013799.1 | 1756 | Intron | XP_016869288.1 | |||
XM_017013800.1 | 1756 | Intron | XP_016869289.1 |