Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGTCTGTTTCTTCCTCTGCATT[A/G]TAAGCTCCTTGGGTGTGGAACCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 184420 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FDFT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FDFT1 - farnesyl-diphosphate farnesyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287742.1 | Intron | NP_001274671.1 | ||||
NM_001287743.1 | Intron | NP_001274672.1 | ||||
NM_001287744.1 | Intron | NP_001274673.1 | ||||
NM_001287745.1 | Intron | NP_001274674.1 | ||||
NM_001287747.1 | Intron | NP_001274676.1 | ||||
NM_001287748.1 | Intron | NP_001274677.1 | ||||
NM_001287749.1 | Intron | NP_001274678.1 | ||||
NM_001287750.1 | Intron | NP_001274679.1 | ||||
NM_001287751.1 | Intron | NP_001274680.1 | ||||
NM_001287756.1 | Intron | NP_001274685.1 | ||||
NM_004462.4 | Intron | NP_004453.3 | ||||
XM_017013217.1 | Intron | XP_016868706.1 |