Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCCAAGCGCCATGGCCGCTGCCG[C/T]CCGAGCCCGGGTCGCGTACTTGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611083 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADHFE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADHFE1 - alcohol dehydrogenase, iron containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144650.2 | 48 | Missense Mutation | GCC,GTC | A,V 5 | NP_653251.2 |
RRS1 - ribosome biogenesis regulator homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RRS1-AS1 - RRS1 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |