Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGCAACGTCTGCTTCTCGGGGT[C/T]CTCGGTCAGCTTCATTCGTGGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 130592 MIM: 611552 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EEF1D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EEF1D - eukaryotic translation elongation factor 1 delta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MROH6 - maestro heat like repeat family member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAPRT - nicotinate phosphoribosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286829.1 | 1127 | Missense Mutation | AAC,GAC | N,D 411 | NP_001273758.1 | |
NM_145201.5 | 1127 | Missense Mutation | AAC,GAC | N,D 411 | NP_660202.3 | |
XM_017013975.1 | 1127 | Missense Mutation | AAC,GAC | N,D 484 | XP_016869464.1 | |
XM_017013976.1 | 1127 | Missense Mutation | AAC,GAC | N,D 484 | XP_016869465.1 | |
XM_017013977.1 | 1127 | Missense Mutation | AAC,GAC | N,D 384 | XP_016869466.1 | |
XM_017013978.1 | 1127 | Missense Mutation | AAC,GAC | N,D 484 | XP_016869467.1 | |
XM_017013979.1 | 1127 | Missense Mutation | AAC,GAC | N,D 183 | XP_016869468.1 | |
XM_017013980.1 | 1127 | Missense Mutation | AAC,GAC | N,D 183 | XP_016869469.1 |