Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGAGCACAGGTGACCGGAGCCAGG[A/C]GGTGAGGCATGGACTGAGGGCGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138200 MIM: 609172 MIM: 603780 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPT - glutamic--pyruvic transaminase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005309.2 | 945 | Missense Mutation | GAG,GCG | E,A 11 | NP_005300.1 | |
XM_011516993.2 | 945 | Missense Mutation | GAG,GCG | E,A 11 | XP_011515295.1 |
LOC101928953 - uncharacterized LOC101928953 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MFSD3 - major facilitator superfamily domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R16A - protein phosphatase 1 regulatory subunit 16A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RECQL4 - RecQ like helicase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |