Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGACCCAACAGGCAAGGGGGCAGG[C/T]GGCCCCCGGGGCAGCTCCGAAACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612757 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPIHBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPIHBP1 - glycosylphosphatidylinositol anchored high density lipoprotein binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301772.1 | 540 | Intron | NP_001288701.1 | |||
NM_178172.5 | 540 | Silent Mutation | GGC,GGT | G,G 155 | NP_835466.2 |