Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGATGCGCAAAGGCTCTGGAGGCC[C/T]GGCGCCCCGTGCACCCCCCAGGTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 617007 | ||||||||||||||||||||
Literature Links: |
TRIM35 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRIM35 - tripartite motif containing 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304495.1 | 1529 | UTR 3 | NP_001291424.1 | |||
NM_171982.4 | 1529 | Missense Mutation | AGG,GGG | R,G 473 | NP_741983.2 | |
XM_005273452.3 | 1529 | UTR 3 | XP_005273509.1 | |||
XM_011544453.1 | 1529 | Missense Mutation | AGG,GGG | R,G 323 | XP_011542755.1 |