Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTGTCAATGTAGTCTTCCAGCTC[A/G]CGGACCTGGAACTCCTTCTTGCTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608737 | ||||||||||||||||||||
Literature Links: |
RAB11FIP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAB11FIP1 - RAB11 family interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002814.2 | 3803 | Silent Mutation | CGC,CGT | R,R 1249 | NP_001002814.2 | |
NM_025151.4 | 3803 | Silent Mutation | CGC,CGT | R,R 615 | NP_079427.4 | |
XM_017013869.1 | 3803 | Intron | XP_016869358.1 |