Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTAAACAACCTTATGCCTGAAC[A/G]TCAGCTAATTCTGGAGGAAGTGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF706 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF706 - zinc finger protein 706 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042510.1 | 519 | Silent Mutation | GAC,GAT | D,D 73 | NP_001035975.1 | |
NM_001267708.1 | 519 | Silent Mutation | GAC,GAT | D,D 73 | NP_001254637.1 | |
NM_001267709.1 | 519 | Silent Mutation | GAC,GAT | D,D 73 | NP_001254638.1 | |
NM_016096.3 | 519 | Silent Mutation | GAC,GAT | D,D 73 | NP_057180.1 |