Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTATTGACCCTCTGGCTAAAGAA[A/G]AAGAAAATGTTGGCATAGATATTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602638 MIM: 600899 | ||||||||||||||||||||
Literature Links: |
MCM4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MCM4 - minichromosome maintenance complex component 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005914.3 | 1483 | Missense Mutation | AAA,GAA | K,E 181 | NP_005905.2 | |
NM_182746.2 | 1483 | Missense Mutation | AAA,GAA | K,E 181 | NP_877423.1 |
PRKDC - protein kinase, DNA-activated, catalytic polypeptide | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |