Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTCTTCCCCAAGGAACGGGCCAC[C/G]GCATCGAAGGGCACGGCATACTCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604997 | ||||||||||||||||||||
Literature Links: |
DOK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DOK2 - docking protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317800.1 | 934 | Silent Mutation | GCC,GCG | A,A 151 | NP_001304729.1 | |
NM_003974.3 | 934 | Silent Mutation | GCC,GCG | A,A 305 | NP_003965.2 | |
NM_201349.2 | 934 | Silent Mutation | GCC,GCG | A,A 151 | NP_958728.1 | |
XM_005273680.4 | 934 | Silent Mutation | GCC,GCG | A,A 211 | XP_005273737.1 |