Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTACTTCTTTCGGTTACAGTTC[G/T]TTGCTTGACCTTCCCTTTTTAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605795 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TEX15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TEX15 - testis expressed 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031271.3 | 10568 | UTR 3 | NP_112561.2 | |||
XM_005273575.3 | 10568 | UTR 3 | XP_005273632.1 | |||
XM_005273576.3 | 10568 | UTR 3 | XP_005273633.1 | |||
XM_006716369.3 | 10568 | UTR 3 | XP_006716432.1 | |||
XM_011544588.2 | 10568 | UTR 3 | XP_011542890.1 | |||
XM_011544589.2 | 10568 | UTR 3 | XP_011542891.1 | |||
XM_011544590.2 | 10568 | UTR 3 | XP_011542892.1 | |||
XM_011544591.2 | 10568 | UTR 3 | XP_011542893.1 | |||
XM_011544592.2 | 10568 | UTR 3 | XP_011542894.1 | |||
XM_011544593.2 | 10568 | UTR 3 | XP_011542895.1 | |||
XM_011544594.2 | 10568 | Intron | XP_011542896.1 |