Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCGTAGTTTCTGTAGAAATCTGC[A/G]TATGCCTTCTTTCTTTGATCAGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 124090 MIM: 607817 | ||||||||||||||||||||
Literature Links: |
COX6C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COX6C - cytochrome c oxidase subunit 6C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004374.3 | 554 | Silent Mutation | TAC,TAT | Y,Y 49 | NP_004365.1 | |
XM_017013020.1 | 554 | Intron | XP_016868509.1 |
VPS13B - vacuolar protein sorting 13 homolog B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |