Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGACATACCTTCATCATGACCTC[A/G]TTGTTCAAGTTCTCACTGATGATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608737 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB11FIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAB11FIP1 - RAB11 family interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002814.2 | 3674 | Silent Mutation | AAC,AAT | N,N 1206 | NP_001002814.2 | |
NM_025151.4 | 3674 | Silent Mutation | AAC,AAT | N,N 572 | NP_079427.4 | |
XM_017013869.1 | 3674 | Intron | XP_016869358.1 |