Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGGAGGCTGGTCCATATCTGTAT[C/T]GTACCCCGTATACTGACTGAAGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609562 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPA6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPA6 - carboxypeptidase A6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020361.4 | 1350 | Missense Mutation | CAA,CGA | Q,R 365 | NP_065094.3 | |
XM_011517570.2 | 1350 | Missense Mutation | CAA,CGA | Q,R 217 | XP_011515872.1 | |
XM_017013646.1 | 1350 | Missense Mutation | CAA,CGA | Q,R 217 | XP_016869135.1 | |
XM_017013647.1 | 1350 | Intron | XP_016869136.1 |
LOC102724708 - uncharacterized LOC102724708 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |