Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACTTACTGTGCGAGTGGGACACC[A/C]TTTTCACCCTCTGACAGATATATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604109 MIM: 601658 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HHLA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HHLA1 - HERV-H LTR-associating 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145095.1 | 1535 | Missense Mutation | AGG,ATG | R,M 512 | NP_001138567.1 |
OC90 - otoconin 90 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |