Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTAAGAACCTCTTCTCCGCCCTCA[A/G]CAGAAGGAGCCATGCTGATAATGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605080 | ||||||||||||||||||||
Literature Links: |
CNGB3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNGB3 - cyclic nucleotide gated channel beta 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019098.4 | 2423 | Missense Mutation | GCT,GTT | A,V 792 | NP_061971.3 | |
XM_011517138.2 | 2423 | Missense Mutation | GCT,GTT | A,V 654 | XP_011515440.1 |