Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCAAGGGTGGCCGACCCCCATCA[C/T]TGATGCGGATCAAGACGACATACTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603017 | ||||||||||||||||||||
Literature Links: |
CDH17 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDH17 - cadherin 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144663.1 | 2366 | Missense Mutation | AAT,AGT | N,S 747 | NP_001138135.1 | |
NM_004063.3 | 2366 | Missense Mutation | AAT,AGT | N,S 747 | NP_004054.3 | |
XM_011516790.1 | 2366 | Missense Mutation | AAT,AGT | N,S 667 | XP_011515092.1 |