Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGTTTTTGTAAAAGTGTACTGC[C/T]CCTCAAGCAAAAAAGAAAGTGGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PURG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PURG - purine rich element binding protein G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015508.2 | Intron | NP_001015508.1 | ||||
NM_001323311.1 | Intron | NP_001310240.1 | ||||
NM_001323312.1 | Intron | NP_001310241.1 | ||||
NM_013357.2 | Intron | NP_037489.1 |