Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCACAAAGACCTTGCTTACTGATC[C/T]GCCTTTGATAGATCTTTTCTTCTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604289 | ||||||||||||||||||||
Literature Links: |
RAD54B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAD54B - RAD54 homolog B (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205262.2 | 2476 | Intron | NP_001192191.1 | |||
NM_001205263.1 | 2476 | Missense Mutation | CAG,CGG | Q,R 598 | NP_001192192.1 | |
NM_012415.3 | 2476 | Missense Mutation | CAG,CGG | Q,R 782 | NP_036547.1 |