Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGGACACTCGTGGCAGCCTGTCT[A/G]GCAGCCTGGGCTCTCTGAAAGTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606823 MIM: 607013 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADGRA2 - adhesion G protein-coupled receptor A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032777.9 | 1354 | Intron | NP_116166.9 | |||
XM_005273471.3 | 1354 | Intron | XP_005273528.1 | |||
XM_011544481.2 | 1354 | Intron | XP_011542783.1 | |||
XM_011544482.2 | 1354 | Intron | XP_011542784.1 | |||
XM_011544483.2 | 1354 | Intron | XP_011542785.1 |
BRF2 - BRF2, RNA polymerase III transcription initiation factor 50 kDa subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018310.3 | 1354 | Silent Mutation | GCC,GCT | A,A 408 | NP_060780.2 |