Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAAAGCACGCGTCAATATCACG[C/T]GTAGGAAAAACTAGAAAATTGTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602728 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC25 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC25 - coiled-coil domain containing 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304529.1 | 3148 | UTR 3 | NP_001291458.1 | |||
NM_001304530.1 | 3148 | UTR 3 | NP_001291459.1 | |||
NM_001304532.1 | 3148 | UTR 3 | NP_001291461.1 | |||
NM_018246.2 | 3148 | UTR 3 | NP_060716.2 | |||
XM_011544570.2 | 3148 | UTR 3 | XP_011542872.1 | |||
XM_011544571.2 | 3148 | UTR 3 | XP_011542873.1 |
SCARA3 - scavenger receptor class A member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016240.2 | 3148 | Intron | NP_057324.2 | |||
NM_182826.1 | 3148 | Intron | NP_878185.1 | |||
XM_017013535.1 | 3148 | Intron | XP_016869024.1 | |||
XM_017013536.1 | 3148 | UTR 3 | XP_016869025.1 | |||
XM_017013537.1 | 3148 | Intron | XP_016869026.1 |