Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTACTGTCCCGTTACTCACACA[C/G]ACGTGCTGGATTCAACACCCACGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613468 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASAH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASAH1 - N-acylsphingosine amidohydrolase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127505.2 | 1723 | UTR 3 | NP_001120977.1 | |||
NM_004315.5 | 1723 | UTR 3 | NP_004306.3 | |||
NM_177924.4 | 1723 | UTR 3 | NP_808592.2 | |||
XM_005273504.3 | 1723 | UTR 3 | XP_005273561.1 |