Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCGACCAAAGGTCTTCCCACACT[C/G]GTGGCAGGCGTAGGGCTTGTCGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601262 | ||||||||||||||||||||
Literature Links: |
ZNF16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF16 - zinc finger protein 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029976.2 | 1747 | Missense Mutation | CAG,GAG | Q,E 521 | NP_001025147.2 | |
NM_006958.2 | 1747 | Missense Mutation | CAG,GAG | Q,E 521 | NP_008889.2 | |
XM_005272341.2 | 1747 | Missense Mutation | CAG,GAG | Q,E 521 | XP_005272398.1 | |
XM_005272342.2 | 1747 | Missense Mutation | CAG,GAG | Q,E 381 | XP_005272399.1 | |
XM_011517298.1 | 1747 | Missense Mutation | CAG,GAG | Q,E 521 | XP_011515600.1 |