Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGGTGGTTCCCAATGCCACTTT[A/C]TTGGAGAAACTTTTGGAAAAATACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CRISPLD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRISPLD1 - cysteine rich secretory protein LCCL domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286777.1 | 603 | Intron | NP_001273706.1 | |||
NM_001286778.1 | 603 | Intron | NP_001273707.1 | |||
NM_031461.5 | 603 | Missense Mutation | TTA,TTC | L,F 31 | NP_113649.1 |